ADV465750295 AChemBlock

tert-Butyl (3S,5S)-3,5-dimethylpiperazine-1-carboxylate

  • MDL number MFCD08686694
  • Cas Number : 888327-50-2
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3s-5s-3-5-dimethylpiperazine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13371
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart