ADV465750302 AChemBlock

2-((tert-butoxycarbonyl)amino)-3-(3,3-difluorocyclobutyl)propanoic acid

  • MDL number MFCD28501989
  • Cas Number : 681128-43-8
  • Molecular Weight : 279.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cyclobutanepropanoic-acid-a-1-1-dimethylethoxy-carbonyl-amino-3-3-difluoro.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13384
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart