ADV465750309 AChemBlock

tert-Butyl n-{1-ethenyl-2-oxabicyclo[2.2.2]octan-4-yl}carbamate

  • MDL number MFCD23099503
  • Cas Number : 1416374-98-5
  • Molecular Weight : 253.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-1-ethenyl-2-oxabicyclo-2-2-2-octan-4-yl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13392
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart