ADV465750316 AChemBlock

tert-Butyl 2-oxo-3,6-diazabicyclo[3.1.1]heptane-6-carboxylate

  • MDL number MFCD18793394
  • Cas Number : 1273562-74-5
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-oxo-3-6-diazabicyclo-3-1-1-heptane-6-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13399
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart