ADV465750319 AChemBlock

tert-Butyl N-{[(2S)-4,4-difluoropyrrolidin-2-yl]methyl}carbamate

  • MDL number MFCD22566251
  • Cas Number : 1363384-66-0
  • Molecular Weight : 236.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2s-4-4-difluoropyrrolidin-2-yl-methyl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13402
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart