ADV465750324 AChemBlock

[(2R)-4,4-Difluoro-1-methylpyrrolidin-2-yl]methanol

  • MDL number MFCD23106101
  • Cas Number : 1408002-86-7
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2r-4-4-difluoro-1-methylpyrrolidin-2-yl-methanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13407
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart