ADV465750334 AChemBlock

2-(Pyridazin-3-yl)acetic acid lithium salt

  • MDL number MFCD28952958
  • Cas Number : 1845687-62-8
  • Molecular Weight : 144.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-pyridazin-3-yl-acetic-acid-lithium-salt.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13418
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart