ADV465750344 AChemBlock

tert-Butyl N-{6-azaspiro[3.4]octan-2-yl}carbamate hydrochloride

  • MDL number MFCD27987993
  • Cas Number : 1630906-42-1
  • Molecular Weight : 262.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-6-azaspiro-3-4-octan-2-yl-carbamate-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13430
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart