ADV465750352 AChemBlock

Decernotinib

  • MDL number MFCD27987896
  • Cas Number : 944842-54-0
  • Molecular Weight : 392.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form White powder
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/decernotinib.html

Safety Information

signalword warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13439
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart