ADV465750355 AChemBlock

(1S,3R,4R)-rel-4-(Boc-amino)-3-fluorocyclohexanol

  • MDL number MFCD24850022
  • Cas Number : 1268512-14-6
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1s-3r-4r-rel-4-boc-amino-3-fluorocyclohexanol.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13442
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart