ADV465750359 AChemBlock

(1s,3r,4s)-Rel-4-(Boc-amino)-3-fluorocyclohexanol

  • MDL number MFCD28502768
  • Cas Number : 1788041-38-2
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1s-3r-4s-rel-4-boc-amino-3-fluorocyclohexanol.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13448
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart