ADV465750361 AChemBlock

(1R,3S)-3-Amino-1-(Boc-amino)cyclohexane

  • MDL number MFCD22394043
  • Cas Number : 1259278-17-5
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1r-3s-3-amino-1-boc-amino-cyclohexane.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13450
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart