ADV465750363 AChemBlock

methyl 4-hydroxybicyclo[2.2.1]heptane-1-carboxylate

  • MDL number MFCD28502791
  • Cas Number : 88888-31-7
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-hydroxybicyclo-2-2-1-heptane-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13452
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart