ADV465750364 AChemBlock

tert-Butyl N-[3-(trifluoromethyl)piperidin-3-yl]carbamate

  • MDL number MFCD23106386
  • Cas Number : 1419101-52-2
  • Molecular Weight : 268.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3-trifluoromethyl-piperidin-3-yl-carbamate.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13453
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart