ADV465750376 AChemBlock

Ethyl 6-bromo-1H-pyrrolo[3,2-c]pyridine-2-carboxylate

  • MDL number MFCD28501562
  • Cas Number : 1788041-67-7
  • Molecular Weight : 269.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-6-bromo-1h-pyrrolo-3-2-c-pyridine-2-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13466
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart