ADV465750379 AChemBlock

tert-Butyl N-{[(2R)-5,5-difluoropiperidin-2-yl]methyl}carbamate

  • MDL number MFCD27987658
  • Cas Number : 1638744-23-6
  • Molecular Weight : 250.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-2r-5-5-difluoropiperidin-2-yl-methyl-carbamate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13470
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart