ADV465750421 AChemBlock

1-[(2-Fluorophenyl)methyl]-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile

  • MDL number MFCD20268231
  • Cas Number : 256376-65-5
  • Molecular Weight : 252.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-2-fluorophenyl-methyl-1h-pyrazolo-3-4-b-pyridine-3-carbonitrile.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13501
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart