ADV465750423 AChemBlock

1-Benzyl-N,4-dimethylpiperidin-3-amine

  • MDL number MFCD12925759
  • Cas Number : 384338-23-2
  • Molecular Weight : 218.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-benzyl-n-4-dimethylpiperidin-3-amine.html

Safety Information

signalword warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13503
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart