ADV465750432 AChemBlock

tert-Butyl 4-(4-amino-1H-pyrazol-1-yl)piperidine-1-carboxylate

  • MDL number MFCD10687120
  • Cas Number : 1029413-55-5
  • Molecular Weight : 266.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-4-amino-1h-pyrazol-1-yl-piperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13512
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart