ADV465750435 AChemBlock

(3S)-N,N-dimethylpyrrolidin-3-amine dihydrochloride

  • MDL number MFCD08561117
  • Cas Number : 144043-20-9
  • Molecular Weight : 187.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3s-n-n-dimethylpyrrolidin-3-amine-dihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13515
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart