ADV465750439 AChemBlock

(3R,5S)-5-(hydroxymethyl)pyrrolidin-3-ol hydrochloride

  • MDL number MFCD09056830
  • Cas Number : 478922-47-3
  • Molecular Weight : 153.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3r-5s-5-hydroxymethyl-pyrrolidin-3-ol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13519
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart