ADV465750444 AChemBlock

4-Bromo-1H,2H,3H-pyrrolo[2,3-b]pyridin-2-one

  • MDL number MFCD12406103
  • Cas Number : 1086064-49-4
  • Molecular Weight : 213.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1h-2h-3h-pyrrolo-2-3-b-pyridin-2-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13524
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart