ADV465750451 AChemBlock

tert-Butyl 4-(6-aminopyridin-3-yl)piperazine-1-carboxylate

  • MDL number MFCD11594962
  • Cas Number : 571188-59-5
  • Molecular Weight : 278.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-6-aminopyridin-3-yl-piperazine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13531
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart