ADV465750458 AChemBlock

1-[(Benzyloxy)carbonyl]-3-methylpiperidine-3-carboxylic acid

  • MDL number MFCD11559118
  • Cas Number : 174543-78-3
  • Molecular Weight : 277.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-benzyloxy-carbonyl-3-methylpiperidine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13538
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart