ADV465750461 AChemBlock

Methyl (3S)-5-oxo-1-[(1R)-1-phenylethyl]pyrrolidine-3-carboxylate

  • MDL number MFCD20526323
  • Cas Number : 99735-46-3
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-3s-5-oxo-1-1r-1-phenylethyl-pyrrolidine-3-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13541
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart