ADV465750462 AChemBlock

Methyl 1-(propan-2-yl)-2,3-dihydro-1H-indole-6-carboxylate

  • MDL number MFCD28501558
  • Cas Number : 1788044-05-2
  • Molecular Weight : 219.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-1-propan-2-yl-2-3-dihydro-1h-indole-6-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13542
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart