ADV465750465 AChemBlock

Oxalic acid bis(methyl 4-{[(tert-butoxy)carbonyl]amino}piperidine-4-carboxylate)

  • MDL number MFCD28502518
  • Cas Number : 1788043-91-3
  • Molecular Weight : 606.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/oxalic-acid-bis-methyl-4-tert-butoxy-carbonyl-amino-piperidine-4-carboxylate.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13545
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart