ADV465750472 AChemBlock

1-(5-Bromo-3-iodo-6-methyl-1H-indazol-1-yl)ethan-1-one

  • MDL number MFCD28501546
  • Cas Number : 1788054-76-1
  • Molecular Weight : 379

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-5-bromo-3-iodo-6-methyl-1h-indazol-1-yl-ethan-1-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13552
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart