ADV465750476 AChemBlock

4-(Hydrazinylmethyl)pyridine trihydrochloride

  • MDL number MFCD20233302
  • Cas Number : 1349717-75-4
  • Molecular Weight : 232.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-hydrazinylmethyl-pyridine-trihydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13556
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart