ADV465750484 AChemBlock

tert-Butyl (2S)-2-(1H-imidazol-2-yl)pyrrolidine-1-carboxylate

  • MDL number MFCD16660093
  • Cas Number : 1007882-58-7
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2s-2-1h-imidazol-2-yl-pyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13564
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart