ADV465750487 AChemBlock

Benzyl (3R)-3-aminopiperidine-1-carboxylate

  • MDL number MFCD05864625
  • Cas Number : 1044560-96-4
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/benzyl-3r-3-aminopiperidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13567
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart