ADV465750489 AChemBlock

4-Chloro-3-iodo-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD11559017
  • Cas Number : 1082950-96-6
  • Molecular Weight : 280.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Light yellow solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-3-iodo-1h-pyrazolo-3-4-d-pyrimidine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13569
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart