ADV465750490 AChemBlock

tert-Butyl 2-methylidene-7-azaspiro[3.5]nonane-7-carboxylate

  • MDL number MFCD28502553
  • Cas Number : 1356476-38-4
  • Molecular Weight : 237.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-2-methylidene-7-azaspiro-3-5-nonane-7-carboxylate.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13580
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart