ADV465750500 AChemBlock

5-Bromo-7-methyl-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD27987377
  • Cas Number : 1638761-56-4
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-7-methyl-7h-pyrrolo-2-3-d-pyrimidine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13590
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart