ADV465750501 AChemBlock

Cyclopropanol

  • MDL number MFCD19707103
  • Cas Number : 16545-68-9
  • Molecular Weight : 58.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form Yellow liquid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/cyclopropanol.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13591
pictograms GHS02
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart