ADV465750503 AChemBlock

N-Boc-cis-4-amino-l-proline methyl ester hydrochloride

  • MDL number MFCD03787927
  • Cas Number : 171110-72-8
  • Molecular Weight : 280.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/n-boc-cis-4-amino-l-proline-methyl-ester-hydrochloride.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13593
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart