ADV465750504 AChemBlock

tert-Butyl n-[(3R)-pyrrolidin-3-ylmethyl]carbamate

  • MDL number MFCD06796615
  • Cas Number : 173340-25-5
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Physical Form White solid
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-n-3r-pyrrolidin-3-ylmethyl-carbamate.html

Safety Information

signalword Warning
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13594
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart