ADV465750506 AChemBlock

4-(Dimethoxymethyl)-2-(methylsulfanyl)pyrimidine

  • MDL number MFCD08275990
  • Cas Number : 180869-36-7
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/4-dimethoxymethyl-2-methylsulfanyl-pyrimidine.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13596
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart