ADV465750510 AChemBlock

Ethyl 1-methyl-3-oxocyclobutane-1-carboxylate

  • MDL number MFCD23105920
  • Cas Number : 227607-44-5
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/ethyl-1-methyl-3-oxocyclobutane-1-carboxylate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13600
pictograms GHS02
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart