ADV465750516 AChemBlock

2-Sulfanylidene-1H,2H,3H,4H-pyrido[3,4-d]pyrimidin-4-one

  • MDL number MFCD07437913
  • Cas Number : 412341-42-5
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-sulfanylidene-1h-2h-3h-4h-pyrido-3-4-d-pyrimidin-4-one.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13606
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart