ADV465750531 AChemBlock

tert-Butyl 4-(1-ethoxycyclopropanecarbonyl)piperazine-1-carboxylate

  • MDL number MFCD28502551
  • Cas Number : 1788041-42-8
  • Molecular Weight : 298.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-1-ethoxycyclopropanecarbonyl-piperazine-1-carboxylate.html

Safety Information

signalword warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13621
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart