ADV465750540 AChemBlock

3-Iodo-6-methyl-1H-indazole

  • MDL number MFCD07781627
  • Cas Number : 885518-96-7
  • Molecular Weight : 258.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/3-iodo-6-methyl-1h-indazole.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13630
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart