ADV465750556 AChemBlock

1H-Pyrazole-3,4-diamine sulfate

  • MDL number MFCD28501207
  • Cas Number : 16258-80-3
  • Molecular Weight : 196.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1h-pyrazole-3-4-diamine-sulfate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13646
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart