ADV465750569 AChemBlock

Methyl 2-(5-bromopyrimidin-2-yl)acetate

  • MDL number MFCD16987768
  • Cas Number : 948594-80-7
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/methyl-2-5-bromopyrimidin-2-yl-acetate.html

Safety Information

signalword Danger
pcodes P201;P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13659
pictograms GHS06
hstatements H301;H319; H331;H401
Availability details for
Quantity
  • Availability
  • Add to Cart