ADV465750575 AChemBlock

(R)-3-Boc-4-ethyl-2,2-dioxo-[1,2,3]oxathiazolidine

  • MDL number MFCD28502421
  • Cas Number : 1417287-40-1
  • Molecular Weight : 251.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/r-3-boc-4-ethyl-2-2-dioxo-1-2-3-oxathiazolidine.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13665
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart