ADV465750576 AChemBlock

2-Oxa-8-azaspiro[4.5]decane oxlate

  • MDL number MFCD24847828
  • Cas Number : 1588441-03-5
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-oxa-8-azaspiro-4-5-decane-oxlate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13666
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart