ADV465750578 AChemBlock

benzyl 3-carbamoyl-3-methylpiperidine-1-carboxylate

  • MDL number MFCD28502766
  • Cas Number : 174543-79-4
  • Molecular Weight : 276.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/1-piperidinecarboxylic-acid-3-aminocarbonyl-3-methyl-phenylmethyl-ester.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13668
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart