ADV465750580 AChemBlock

2-{[(tert-Butoxy)carbonyl]amino}-3-cyclobutylpropanoic acid

  • MDL number MFCD09264344
  • Cas Number : 565456-75-9
  • Molecular Weight : 243.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-tert-butoxy-carbonyl-amino-3-cyclobutylpropanoic-acid.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13670
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart