ADV465750583 AChemBlock

tert-Butyl (3R)-3-({[(benzyloxy)carbonyl]amino}methyl)pyrrolidine-1-carboxylate

  • MDL number MFCD19704307
  • Cas Number : 879275-54-4
  • Molecular Weight : 334.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-3r-3-benzyloxy-carbonyl-amino-methyl-pyrrolidine-1-carboxylate.html

Safety Information

signalword Warning
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13673
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart