ADV465750587 AChemBlock

2-(Methylsulfanyl)pyrimidine-4-carbaldehyde

  • MDL number MFCD02683211
  • Cas Number : 1074-68-6
  • Molecular Weight : 154.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Shipping Temperature Store at 0-8
Country of Origin China
Product Link https://www.achemblock.com/2-methylsulfanyl-pyrimidine-4-carbaldehyde.html

Safety Information

signalword WARNING
pcodes P261;P305+351+338;P302+352
MSDS https://www.achemblock.com/AC_SDS.php?Catalog=L13677
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart